Merck
  • A microwave synthesis of mesoporous NiCo2O4 nanosheets as electrode materials for lithium-ion batteries and supercapacitors.
A microwave synthesis of mesoporous NiCo2O4 nanosheets as electrode materials for lithium-ion batteries and supercapacitors.
Chemphyschem : a European journal of chemical physics and physical chemistry (2014-11-06)
Anjon Kumar Mondal, Dawei Su, Shuangqiang Chen, Katja Kretschmer, Xiuqiang Xie, Hyo-Jun Ahn, Guoxiu Wang
ABSTRACT
A facile microwave method was employed to synthesize NiCo2 O4 nanosheets as electrode materials for lithium-ion batteries and supercapacitors. The structure and morphology of the materials were characterized by X-ray diffraction, field-emission scanning electron microscopy, transmission electron microscopy and Brunauer-Emmett-Teller methods. Owing to the porous nanosheet structure, the NiCo2 O4 electrodes exhibited a high reversible capacity of 891 mA h g(-1) at a current density of 100 mA g(-1) , good rate capability and stable cycling performance. When used as electrode materials for supercapacitors, NiCo2 O4 nanosheets demonstrated a specific capacitance of 400 F g(-1) at a current density of 20 A g(-1) and superior cycling stability over 5000 cycles. The excellent electrochemical performance could be ascribed to the thin porous structure of the nanosheets, which provides a high specific surface area to increase the electrode-electrolyte contact area and facilitate rapid ion transport.
Sigma-Aldrich
Nickel(II) nitrate hexahydrate, ≥99.9% trace metals basis