Merck
  • Lipid-polymer hybrid nanoparticles as a new generation therapeutic delivery platform: a review.
Lipid-polymer hybrid nanoparticles as a new generation therapeutic delivery platform: a review.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2013-07-23)
Kunn Hadinoto, Ajitha Sundaresan, Wean Sin Cheow
ABSTRACT
Lipid-polymer hybrid nanoparticles (LPNs) are core-shell nanoparticle structures comprising polymer cores and lipid/lipid-PEG shells, which exhibit complementary characteristics of both polymeric nanoparticles and liposomes, particularly in terms of their physical stability and biocompatibility. Significantly, the LPNs have recently been demonstrated to exhibit superior in vivo cellular delivery efficacy compared to that obtained from polymeric nanoparticles and liposomes. Since their inception, the LPNs have advanced significantly in terms of their preparation strategy and scope of applications. Their preparation strategy has undergone a shift from the conceptually simple two-step method, involving preformed polymeric nanoparticles and lipid vesicles, to the more principally complex, yet easier to perform, one-step method, relying on simultaneous self-assembly of the lipid and polymer, which has resulted in better products and higher production throughput. The scope of LPNs' applications has also been extended beyond single drug delivery for anticancer therapy, to include combinatorial and active targeted drug deliveries, and deliveries of genetic materials, vaccines, and diagnostic imaging agents. This review details the current state of development for the LPNs preparation and applications from which we identify future research works needed to bring the LPNs closer to its clinical realization.
Sigma-Aldrich
Folic acid modified polyethylenimine solution, high molecular weight, 10 mg/ml aqueous solution, suitable for biomedical research
Sigma-Aldrich
Folic acid modified polyethylenimine solution, low molecular weight, 10 mg/mL (aqueous solution), suitable for biomedical research