Merck
  • RNA helicase DDX3 regulates RAD51 localization and DNA damage repair in Ewing sarcoma.
RNA helicase DDX3 regulates RAD51 localization and DNA damage repair in Ewing sarcoma.
iScience (2024-02-07)
Matthew E Randolph, Marwa Afifi, Aparna Gorthi, Rachel Weil, Breelyn A Wilky, Joshua Weinreb, Paul Ciero, Natalie Ter Hoeve, Paul J van Diest, Venu Raman, Alexander J R Bishop, David M Loeb
ABSTRACT
We previously demonstrated that RNA helicase DDX3X (DDX3) can be a therapeutic target in Ewing sarcoma (EWS), but its role in EWS biology remains unclear. The present work demonstrates that DDX3 plays a unique role in DNA damage repair (DDR). We show that DDX3 interacts with several proteins involved in homologous recombination, including RAD51, RECQL1, RPA32, and XRCC2. In particular, DDX3 colocalizes with RAD51 and RNA:DNA hybrid structures in the cytoplasm of EWS cells. Inhibition of DDX3 RNA helicase activity increases cytoplasmic RNA:DNA hybrids, sequestering RAD51 in the cytoplasm, which impairs nuclear translocation of RAD51 to sites of double-stranded DNA breaks, thus increasing sensitivity of EWS to radiation treatment, both in vitro and in vivo. This discovery lays the foundation for exploring new therapeutic approaches directed at manipulating DDR protein localization in solid tumors.
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid
Sigma-Aldrich
Anti-DNA Antibody, single stranded, clone TNT-3, Chemicon®, from mouse
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse