Merck
  • Residues in the 11 A channel of histone deacetylase 1 promote catalytic activity: implications for designing isoform-selective histone deacetylase inhibitors.
Residues in the 11 A channel of histone deacetylase 1 promote catalytic activity: implications for designing isoform-selective histone deacetylase inhibitors.
Journal of medicinal chemistry (2008-08-30)
Sujith V W Weerasinghe, Guillermina Estiu, Olaf Wiest, Mary Kay H Pflum
ABSTRACT
Histone deacetylase 1 (HDAC1) has been linked to cell growth and cell cycle regulation, which makes it a widely recognized target for anticancer drugs. Whereas variations of the metal-binding and capping groups of HDAC inhibitors have been studied extensively, the role of the linker region is less well known, despite the potency of inhibitors with diverse linkers, such as MS-275. To facilitate a drug design that targets HDAC1, we assessed the influence of residues in the 11 A channel of the HDAC1 active site on activity by using an alanine scan. The mutation of eight channel residues to alanine resulted in a substantial reduction in deacetylase activity. Molecular dynamics simulations indicated that alanine mutation results in significant movement of the active-site channel, which suggests that channel residues promote HDAC1 activity by influencing substrate interactions. With little characterization of HDAC1 available, the combined experimental and computational results define the active-site residues of HDAC1 that are critical for substrate/inhibitor binding and provide important insight into drug design.
Sigma-Aldrich
Agarose, Low EEO, for Immunoelectrophoresis
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature
Sigma-Aldrich
Agarose, High Gelling Temperature
Sigma-Aldrich
Agarose, Special High EEO
Sigma-Aldrich
Agarose, BioReagent, Molecular Biology, low EEO
Sigma-Aldrich
Agarose, For pulsed field electrophoresis running gel
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature, molecular biology grade
Sigma-Aldrich
Agarose, Type I-A, low EEO
Sigma-Aldrich
Agarose, Type I, low EEO
Sigma-Aldrich
Agarose, Medium EEO, Molecular Biology
Sigma-Aldrich
Agarose, Low EEO
Sigma-Aldrich
Agarose, Molecular Biology
Sigma-Aldrich
Agarose, Type II-A, Medium EEO
Sigma-Aldrich
Agarose, Type IV, Special High EEO
Sigma-Aldrich
Agarose, Wide range, Molecular Biology
Sigma-Aldrich
Agarose, BioReagent, Molecular Biology, Wide range/Standard 3:1
Sigma-Aldrich
Agarose, High EEO
Sigma-Aldrich
Agarose, High EEO