Merck
  • The Alzheimer's disease-linked protease BACE2 cleaves VEGFR3 and modulates its signaling.
The Alzheimer's disease-linked protease BACE2 cleaves VEGFR3 and modulates its signaling.
The Journal of clinical investigation (2024-06-18)
Andree Schmidt, Brian Hrupka, Frauke van Bebber, Sanjay Sunil Kumar, Xiao Feng, Sarah K Tschirner, Marlene Aßfalg, Stephan A Müller, Laura Sophie Hilger, Laura I Hofmann, Martina Pigoni, Georg Jocher, Iryna Voytyuk, Emily L Self, Mana Ito, Kana Hyakkoku, Akimasa Yoshimura, Naotaka Horiguchi, Regina Feederle, Bart De Strooper, Stefan Schulte-Merker, Eckhard Lammert, Dieder Moechars, Bettina Schmid, Stefan F Lichtenthaler
ABSTRACT
The β-secretase β-site APP cleaving enzyme (BACE1) is a central drug target for Alzheimer's disease. Clinically tested, BACE1-directed inhibitors also block the homologous protease BACE2. Yet little is known about physiological BACE2 substrates and functions in vivo. Here, we identify BACE2 as the protease shedding the lymphangiogenic vascular endothelial growth factor receptor 3 (VEGFR3). Inactivation of BACE2, but not BACE1, inhibited shedding of VEGFR3 from primary human lymphatic endothelial cells (LECs) and reduced release of the shed, soluble VEGFR3 (sVEGFR3) ectodomain into the blood of mice, nonhuman primates, and humans. Functionally, BACE2 inactivation increased full-length VEGFR3 and enhanced VEGFR3 signaling in LECs and also in vivo in zebrafish, where enhanced migration of LECs was observed. Thus, this study identifies BACE2 as a modulator of lymphangiogenic VEGFR3 signaling and demonstrates the utility of sVEGFR3 as a pharmacodynamic plasma marker for BACE2 activity in vivo, a prerequisite for developing BACE1-selective inhibitors for safer prevention of Alzheimer's disease.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
β-Secretase Inhibitor IV, β-Secretase Inhibitor IV, CAS 797035-11-1, is a cell-permeable inhibitor that binds to BACE-1 active site and blocks its proteolytic activity (IC50 = 15 nM for human BACE-1).