Merck
  • The interferon γ pathway enhances pluripotency and X-chromosome reactivation in iPSC reprogramming.
The interferon γ pathway enhances pluripotency and X-chromosome reactivation in iPSC reprogramming.
Science advances (2024-08-07)
Mercedes Barrero, Aleksey Lazarenkov, Enrique Blanco, Luis G Palma, Anna V López-Rubio, Moritz Bauer, Anna Bigas, Luciano Di Croce, José Luis Sardina, Bernhard Payer
ABSTRACT
Reprogramming somatic cells into induced pluripotent stem cells (iPSCs) requires activation of the pluripotency network and resetting of the epigenome by erasing the epigenetic memory of the somatic state. In female mouse cells, a critical epigenetic reprogramming step is the reactivation of the inactive X chromosome. Despite its importance, a systematic understanding of the regulatory networks linking pluripotency and X-reactivation is missing. Here, we reveal important pathways for pluripotency acquisition and X-reactivation using a genome-wide CRISPR screen during neural precursor to iPSC reprogramming. In particular, we discover that activation of the interferon γ (IFNγ) pathway early during reprogramming accelerates pluripotency acquisition and X-reactivation. IFNγ stimulates STAT3 signaling and the pluripotency network and leads to enhanced TET-mediated DNA demethylation, which consequently boosts X-reactivation. We therefore gain a mechanistic understanding of the role of IFNγ in reprogramming and X-reactivation and provide a comprehensive resource of the molecular networks involved in these processes.
Sigma-Aldrich
Anti-Stage-Specific Embryonic Antigen-1 Antibody, clone MC-480, clone MC-480, Chemicon®, from mouse
Sigma-Aldrich
CHIR99021, ≥98% (HPLC)
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture