Merck
  • Dynamics of mitochondrial [Ca(2+)] measured with the low-Ca(2+)-affinity dye rhod-5N.
Dynamics of mitochondrial [Ca(2+)] measured with the low-Ca(2+)-affinity dye rhod-5N.
Cell calcium (2011-12-03)
Sergio de la Fuente, Rosalba I Fonteriz, Mayte Montero, Javier Alvarez
ABSTRACT
Available methods to measure mitochondrial [Ca(2+)] ([Ca(2+)](M)) include both targeted proteins and fluorescent dyes. Targeted proteins usually report much higher [Ca(2+)](M) values than fluorescent dyes, up to two orders of magnitude. However, we show here that the low-Ca(2+)-affinity dye rhod-5N provides [Ca(2+)](M) values similar to those reported by targeted aequorin, suggesting that the discrepancies are mainly due to the higher Ca(2+)-affinity of the fluorescent dyes used. We find rhod-5N has an apparent in situ intramitochondrial Kd around 0.5mM. Addition of Ca(2+) buffers containing between 4.5 and 10μM [Ca(2+)] to permeabilized cells loaded with rhod-5N induced increases in calibrated [Ca(2+)](M) up to the 100μM-1mM range, which were dependent on mitochondrial membrane potential. Ca(2+) release from mitochondria was largely dependent on [Na(+)]. We have then used rhod-5N loaded cells to investigate the [Ca(2+)](M) response to agonist stimulation at the single-cell and subcellular level. The [Ca(2+)](M) peaks induced by histamine varied by nearly 10-fold among different cells, with a mean about 25μM. In the presence of the Ca(2+) uniporter stimulator kaempferol, the [Ca(2+)](M) peaks induced by histamine were also highly variable, and the mean [Ca(2+)](M) peak was 3-fold higher. Simultaneous measurement of cytosolic and mitochondrial [Ca(2+)] peaks showed little correlation among the heights of the peaks in both compartments. Studying the [Ca(2+)](M) peaks at the subcellular level, we found significant heterogeneities among regions in the same cell. In particular, the [Ca(2+)](M) increase in mitochondrial regions close to the nucleus was more than double that of mitochondrial regions far from the nucleus.
Product Description
Rhod-5N potassium salt, ≥95% (HPLC)