Merck
  • Growth kinetics and toxicity of Enterobacter cloacae grown on linear alkylbenzene sulfonate as sole carbon source.
Growth kinetics and toxicity of Enterobacter cloacae grown on linear alkylbenzene sulfonate as sole carbon source.
Current microbiology (2008-07-30)
Khaled M Khleifat, Khaled A Tarawneh, Mohammad Ali Wedyan, Amjad A Al-Tarawneh, Khalid Al Sharafa
ABSTRACT
A successful attempt was made to isolate linear alkylbenzene sulfonate (LAS)-degrading bacteria from soil irrigated with wastewater. The isolated bacteria were able to use LAS as sole carbon and energy source. Maximum growth rates on LAS reached only 0.27 h(-1). 16S-rRNA sequencing and fatty-acid analysis placed the bacteria in the genus Enterobacter cloacae. The growth curves of E. cloacae both in the presence of and the absence of LAS were monitored using measurements of optical density at 600 nm in two different media, nutrient broth and M9 minimal medium, and were modeled mathematically. Growth in NB fit the Riccati and Voltera models, indicating that LAS is not toxic to E. cloacae cells. However, growth of E. cloacae in LAS-containing MM fit the Riccati and Voltera models, whereas growth in LAS-free MM fit the Riccati model only. Furthermore, the kinetic data shown were modeled by Monod's, Andrew's, and Tessier's specific growth rate equations, coupled with the rate of consumption of different concentrations of LAS as sole carbon and energy source, and we determined that Andrew's model best fit these data adequately as a result of the cell-inhibitory effect.
Product Description
Millipore
M9 Broth, NutriSelect® Basic, powder, for coliforms, for Escherichia coli, pack of 500 g, suitable for microbiology