Merck
  • Induction of DNA damage signaling genes in benzidine-treated HepG2 cells.
Induction of DNA damage signaling genes in benzidine-treated HepG2 cells.
Environmental and molecular mutagenesis (2011-08-06)
Ssu Ching Chen, You-Cheng Hseu, Jia-Chuen Sung, Chin-Hui Chen, Lei-Chin Chen, King-Tom Chung
ABSTRACT
We examined genotoxicity and DNA damage response in HepG2 cells following exposure to benzidine. Using the Comet assay, we showed that benzidine (50-200 μM) induces DNA damage in HepG2 cells. DNA damage signaling pathway-based PCR arrays were used to investigate expression changes in genes involved in cell-cycle arrest, apoptosis, and DNA repair and showed upregulation of 23 genes and downregulation of one gene in benzidine-treated cells. Induction of G2/M arrest and apoptosis was confirmed at the protein level. Real-time PCR and Western blots were used to demonstrate the expression of select DNA repair-associated genes from the PCR array. Upregulation of the p53 protein in benzidine-treated cells suggests the induction of the p53 DNA damage signaling pathway. Collectively, DNA damage response genes induced by benzidine indicate recruitment complex molecular machinery involved in DNA repair, cell-cycle arrest, and potentially, activation of the apoptosis.
Sigma-Aldrich
Benzidine, ≥98%
Sigma-Aldrich
Benzidine dihydrochloride, ≥99% (titration)
Supelco
Benzidine, analytical standard