Merck
  • Oral administration of a small molecule targeted to block proNGF binding to p75 promotes myelin sparing and functional recovery after spinal cord injury.
Oral administration of a small molecule targeted to block proNGF binding to p75 promotes myelin sparing and functional recovery after spinal cord injury.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2013-01-11)
Chhavy Tep, Tae Hee Lim, Pyung On Ko, Sami Getahun, Jae Cheon Ryu, Virginia M Goettl, Stephen M Massa, Michele Basso, Frank M Longo, Sung Ok Yoon
ABSTRACT
The lack of effective therapies for spinal cord injury points to the need for identifying novel targets for therapeutic intervention. Here we report that a small molecule, LM11A-31, developed to block proNGF-p75 interaction and p75-mediated cell death crosses the blood-brain barrier efficiently when delivered orally. Administered starting 4 h postinjury, LM11A-31 promotes functional recovery without causing any toxicity or increased pain in a mouse model of spinal contusion injury. In both weight-bearing open-field tests and nonweight-bearing swim tests, LM11A-31 was effective in improving motor function and coordination. Such functional improvement correlated with a >50% increase in the number of surviving oligodendrocytes and myelinated axons. We also demonstrate that LM11A-31 indeed inhibits proNGF-p75 interaction in vivo, thereby curtailing the JNK3-mediated apoptotic cascade. These results thus highlight p75 as a novel therapeutic target for an orally delivered treatment for spinal cord injury.
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
Supelco
L-Isoleucine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
L-Isoleucine
Supelco
L-Isoleucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Isoleucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Isoleucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Isoleucine, reagent grade, ≥98% (HPLC)