Merck
  • Biologically different subgroups of invasive ductal carcinoma of the pancreas: Dpc4 status according to the ratio of intraductal carcinoma components.
Biologically different subgroups of invasive ductal carcinoma of the pancreas: Dpc4 status according to the ratio of intraductal carcinoma components.
Clinical cancer research : an official journal of the American Association for Cancer Research (2004-06-03)
Hidenori Takahashi, Tatsuya Oda, Takahiro Hasebe, Yasuyuki Aoyagi, Taira Kinoshita, Masaru Konishi, Toshio Nakagohri, Kazuto Inoue, Shinichiro Takahashi, Hiroshi Kawahira, Morito Monden, Atsushi Ochiai
ABSTRACT
Among invasive ductal carcinomas of the pancreas (IDCP), there is a morphologically characteristic subgroup accompanied by abundant intraductal carcinoma components (ICCs). With the aim of determining whether ICC-rich IDCP are biologically different from ICC-poor IDCP, the expression status of Dpc4 protein was analyzed. A total of 43 IDCP was subdivided into two groups: (a). ICC-rich IDCP (ICCs area occupies >or=10% of the entire tumorous area); and (b). ICC-poor IDCP (with <10% of ICCs area). A total of 10 invasive carcinomas derived from intraductal papillary-mucinous neoplasms (ICs from IPMNs) were also analyzed. Each invasive and intraductal carcinoma area was then evaluated for Dpc4 protein status by immunohistochemistry. In a total of 43 IDCP, there were 23 ICC-rich IDCP and 20 ICC-poor IDCP. Dpc4-positive immunostaining was observed in the invasive carcinoma component of ICC-rich IDCP, ICC-poor IDCP, and ICs from IPMN in 18 of 23 (78%), 4 of 20 (20%), and 7 of 10 (70%) cases, respectively. In the intraductal component, positive staining for Dpc4 was found in 20 of 23 (87%), 3 of 7 (41%), and 8 of 10 (80%) cases, respectively. Dpc4 expression was found in both the invasive and ICC components of ICC-rich IDCP, similar to that found in IC derived from IPMN, whereas the expression of Dpc4 was largely diminished in ICC-poor IDCP. Morphologically distinct subgroups of invasive ductal carcinomas of the pancreas, namely ICC-rich IDCP and ICC-poor IDCP, are also biologically distinguishable as revealed by the differential expression of Dpc4.