Merck
Current topics on inhibitors of respiratory complex I.
Biochimica et biophysica acta (2015-12-03)
Masatoshi Murai, Hideto Miyoshi
ABSTRACT
There are a variety of chemicals which regulate the functions of bacterial and mitochondrial complex I. Some of them, such as rotenone and piericidin A, have been indispensable molecular tools in mechanistic studies on complex I. A large amount of experimental data characterizing the actions of complex I inhibitors has been accumulated so far. Recent X-ray crystallographic structural models of entire complex I may be helpful to carefully interpret this data. We herein focused on recent hot topics on complex I inhibitors and the subjects closely connected to these inhibitors, which may provide useful information not only on the structural and functional aspects of complex I, but also on drug design targeting this enzyme. This article is part of a Special Issue entitled Respiratory complex I, edited by Volker Zickermann and Ulrich Brandt.
Product Description
Piericidin A solution, ≥95% (HPLC), solution, mitochondrial complex I inhibitor