Merck
  • Human intestinal disaccharidases and hereditary disaccharide intolerance. The hydrolysis of sucrose, isomaltose, palatinose (isomaltulose), and a 1,6-alpha-oligosaccharide (isomalto-oligosaccharide) preparation.
Human intestinal disaccharidases and hereditary disaccharide intolerance. The hydrolysis of sucrose, isomaltose, palatinose (isomaltulose), and a 1,6-alpha-oligosaccharide (isomalto-oligosaccharide) preparation.
The Journal of clinical investigation (1963-04-01)
A DAHLQVIST, S AURICCHIO, G SEMENZA, A PRADER
Sigma-Aldrich
Palatinose hydrate, ≥99% (GC)