  • Search Results
  • NLRP3 inflammasome activation in murine alveolar macrophages and related lung pathology is associated with MWCNT nickel contamination.
NLRP3 inflammasome activation in murine alveolar macrophages and related lung pathology is associated with MWCNT nickel contamination.
Inhalation toxicology (2012-12-12)
Raymond F Hamilton, Mary Buford, Chengcheng Xiang, Nianqiang Wu, Andrij Holian
ABSTRACT
Multi-walled carbon nanotubes (MWCNT) have been reported to cause lung pathologies in multiple studies. However, the mechanism responsible for the bioactivity has not been determined. This study used nine different well-characterized MWCNT and examined the outcomes in vitro and in vivo. MWCNT, from a variety of sources that differed primarily in overall purity and metal contaminants, were examined for their effects in vitro (toxicity and NLRP3 inflammasome activation using primary alveolar macrophages isolated from C57Bl/6 mice). In addition, in vivo exposures were conducted to determine the inflammatory and pathogenic potency. The particles produced a differential magnitude of responses, both in vivo and in vitro, that was associated most strongly with nickel contamination on the particle. Furthermore, the mechanism of action for the Ni-contaminated particles was in their ability to disrupt macrophage phagolysosomes, which resulted in NLRP3 activation and subsequent cytokine release associated with prolonged inflammation and lung pathology.
Sigma-Aldrich
Cathepsin B from human placenta, lyophilized powder, ≥5 units/mg protein
Sigma-Aldrich
Cathepsin B from human liver, buffered aqueous solution, ≥1,500 units/mg protein (E1%/280)
Sigma-Aldrich
Cathepsin B from bovine spleen, lyophilized powder, ≥10 units/mg protein