  • Search Results
  • Preparation and evaluation of a new releasable PEGylated tumor necrosis factor-α (TNF-α) conjugate for therapeutic application.
Preparation and evaluation of a new releasable PEGylated tumor necrosis factor-α (TNF-α) conjugate for therapeutic application.
Science China. Life sciences (2013-01-15)
ChuanYun Dai, Ya Fu, ShaoCheng Chen, Biao Li, Bo Yao, WanHong Liu, LiQing Zhu, Nan Chen, Ji Chen, Qiang Zhang
ABSTRACT
To design a releasable PEGylated TNF-α (rPEG-TNF-α), a cathepsin B-sensitive dipeptide (Val-Cit moiety) was inserted into conventional PEG-modified TNF-α (PEG-TNF-α), facilitating its clinical use for anti-tumor therapy. Comparative pharmacokinetic and pharmacodynamic studies showed that the half-lives of both PEGylated forms of TNF-α were ∼60-fold greater than that of unmodified TNF-α. In addition, the in vitro bioactivity of rPEG-TNF-α was greater than that of PEG-TNF-α with the same degree of PEG modification. Release of TNF-α from rPEG-TNF-α in vitro was dependent on the presence of cathepsin B and was inhibited by a cathepsin B inhibitor. Despite the potent cytotoxicity of unmodified TNF-α against normal cells, its PEGylated forms at higher TNF-α concentrations showed low cytotoxic activity against these cells. In contrast, both forms of PEGylated TNF-α showed potent cytotoxic activity against the B16 and L929 cell lines, with rPEG-TNF-α being 5- and 9-fold more potent, respectively, than PEG-TNF-α. Moreover, rPEG-TNF-α was a more potent in vivo antitumor agent than PEG-TNF-α.
Sigma-Aldrich
Cathepsin B from bovine spleen, lyophilized powder, ≥10 units/mg protein
Sigma-Aldrich
N-[(2S,3R)-3-Amino-2-hydroxy-4-phenylbutyryl]-L-leucine, 97%
Sigma-Aldrich
Cathepsin B from human liver, buffered aqueous solution, ≥1,500 units/mg protein (E1%/280)
Sigma-Aldrich
Cathepsin B from human placenta, lyophilized powder, ≥5 units/mg protein
Sigma-Aldrich
Bestatin hydrochloride, ≥98% (HPLC)