  • Search Results
  • Acetate-induced apoptosis in colorectal carcinoma cells involves lysosomal membrane permeabilization and cathepsin D release.
Acetate-induced apoptosis in colorectal carcinoma cells involves lysosomal membrane permeabilization and cathepsin D release.
Cell death & disease (2013-02-23)
C Marques, C S F Oliveira, S Alves, S R Chaves, O P Coutinho, M Côrte-Real, A Preto
ABSTRACT
Colorectal carcinoma (CRC) is one of the most common causes of cancer-related mortality. Short-chain fatty acids secreted by dietary propionibacteria from the intestine, such as acetate, induce apoptosis in CRC cells and may therefore be relevant in CRC prevention and therapy. We previously reported that acetic acid-induced apoptosis in Saccharomyces cerevisiae cells involves partial vacuole permeabilization and release of Pep4p, the yeast cathepsin D (CatD), which has a protective role in this process. In cancer cells, lysosomes have emerged as key players in apoptosis through selective lysosomal membrane permeabilization (LMP) and release of cathepsins. However, the role of CatD in CRC survival is controversial and has not been assessed in response to acetate. We aimed to ascertain whether LMP and CatD are involved in acetate-induced apoptosis in CRC cells. We showed that acetate per se inhibits proliferation and induces apoptosis. More importantly, we uncovered that acetate triggers LMP and CatD release to the cytosol. Pepstatin A (a CatD inhibitor) but not E64d (a cathepsin B and L inhibitor) increased acetate-induced apoptosis of CRC cells, suggesting that CatD has a protective role in this process. Our data indicate that acetate induces LMP and subsequent release of CatD in CRC cells undergoing apoptosis, and suggest exploiting novel strategies using acetate as a prevention/therapeutic agent in CRC, through simultaneous treatment with CatD inhibitors.
Sigma-Aldrich
Cathepsin L from human liver, ≥0.5 units/mg protein, solution
Sigma-Aldrich
Cathepsin B from human placenta, lyophilized powder, ≥5 units/mg protein
Sigma-Aldrich
Cathepsin D from human liver, lyophilized powder, ≥250 units/mg protein (E1%/280)
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
L-Leucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Leucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Cathepsin B from human liver, buffered aqueous solution, ≥1,500 units/mg protein (E1%/280)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
L-Leucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
L-Leucine, 99%, FG
Sigma-Aldrich
Cathepsin D from bovine spleen, lyophilized powder, ≥2.0 units/mg protein
Sigma-Aldrich
Cathepsin B from bovine spleen, lyophilized powder, ≥10 units/mg protein
Supelco
L-Leucine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Supelco
L-Leucine, certified reference material, TraceCERT®
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC