Merck
  • Absence of NF-κB subunit p50 ameliorates cold immobilization stress-induced gastric ulcers.
Absence of NF-κB subunit p50 ameliorates cold immobilization stress-induced gastric ulcers.
Biochemical and biophysical research communications (2013-04-16)
Bin Ye, Pan-Yu Zhou, Meng Jia, Xiao-Song Cheng, Yi-Tao Jia, Shuo-Gui Xu
ABSTRACT
Stress ulcers are a common complication in critically ill patients, but the underlying mechanism is little known. This study characterized the function of the p50 subunit of NF-κB in an experimental model of cold immobilization stress-induced gastric ulcers. Stress-induced gastric mucosal inflammation and gastric injury were examined in wild-type and NF-κB p50-deficient mice. When subjected to cold immobilization stress, NF-κB was rapidly activated in the gastric mucosa in WT mice whereas the majority of κB DNA-binding activity was abrogated from p50(-/-) mice. Deficiency of p50 ameliorated stress-induced expression of TNF-α, MIP-2, and ICAM-1, resulting in reduced mucosal accumulation of neutrophils and gastric injury. These data indicated a critical role for the p50 in the gastric mucosal inflammatory response to cold restraint stress.
Sigma-Aldrich
Peroxidase from horseradish, Type X, ammonium sulfate suspension
Sigma-Aldrich
Peroxidase from horseradish, Type II, essentially salt-free, lyophilized powder, 150-250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Peroxidase from horseradish, Type VI, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Peroxidase from horseradish, Type I, essentially salt-free, lyophilized powder, ≥50 units/mg solid (using pyrogallol)
Sigma-Aldrich
Peroxidase from horseradish, Type XII, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Lactoperoxidase from bovine milk, lyophilized powder (essentially salt-free), ≥200 units/mg protein
Sigma-Aldrich
Peroxidase from horseradish, Type VI-A, essentially salt-free, lyophilized powder, 950-2000 units/mg solid (using ABTS), ≥250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Peroxidase from horseradish, Highly stabilized, essentially salt-free, lyophilized powder, 200-300 units/mg solid (using pyrogallol)
Sigma-Aldrich
Peroxidase from horseradish, lyophilized, powder, ~150 U/mg
Sigma-Aldrich
Lactoperoxidase from bovine milk, lyophilized, powder, ≥150 U/mg
Sigma-Aldrich
Myeloperoxidase from human leukocytes, lyophilized powder, ≥50 units/mg protein