Merck
  • Individual and combined action of pancreatic lipase and pancreatic lipase-related proteins 1 and 2 on native versus homogenized milk fat globules.
Individual and combined action of pancreatic lipase and pancreatic lipase-related proteins 1 and 2 on native versus homogenized milk fat globules.
Molecular nutrition & food research (2009-10-14)
Amélie Berton, Corinne Sebban-Kreuzer, Stéphanie Rouvellac, Christelle Lopez, Isabelle Crenon
ABSTRACT
Pancreatic lipase (PL) and pancreatic lipase-related proteins 1 and 2 (PLRP1 and PLRP2) display different functional properties, despite close structures. The aim of the study was to compare the kinetic properties of recombinant human PLRP1, PLRP2, and PL on a physiological substrate: the milk fat under native and homogenized structures. No lipolytic activity is measured for PLRP1. PLRP2 hydrolyses milk fat with a lower catalytic efficiency than that of PL. PLRP2 activity, higher on homogenized than on native milk fat, is differently influenced by fatty acids (FA) and colipase depending on a proteolytic cleavage in the lid domain. FA enhance the activity on both milks. A colipase positive effect on the non-proteolyzed PLRP2 is observed on homogenized milk and with FA on native milk fat. Bile salts are necessary. An original observation is a synergic effect between PL and PLRP2 on the two milks. An inhibitory effect of PLRP1 on PL activity is also demonstrated. The combined action of pancreatic lipases on milk fat digestion proposes PLRPs as modulators of PL. Our study supports the hypothesis of a major role of PLRP2 in fat digestion in newborns and brings new insights to understand the physiological role of PLRPs.
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger