Merck
  • Detection of hydrazine, methylhydrazine and isoniazid by capillary electrophoresis with a 4-pyridyl hydroquinone self-assembled microdisk platinum electrode.
Detection of hydrazine, methylhydrazine and isoniazid by capillary electrophoresis with a 4-pyridyl hydroquinone self-assembled microdisk platinum electrode.
Journal of pharmaceutical and biomedical analysis (2000-03-04)
T You, L Niu, J Y Gui, S Dong, E Wang
ABSTRACT
Capillary electrophoresis (CE)/electrochemical detection (EC) for the simultaneous detection of hydrazine, methylhydrazine, and isoniazid has been developed with a 4-pyridyl hydroquinone self-assembled microdisk platinum electrode. Such an electrode has very high catalytic ability for hydrazines and they could be detected even at 0.0 V. The responses for hydrazine, methylhydrazine, and isoniazid are linear over 3 orders of detected concentration and of magnitude of 0.2-400 microM, 0.2-400 microM, 0.5 microM-2 mM, with correlation coefficients of 0.9998, 0.9991, and 0.9982, respectively. And they could be detected to levels of 0.1, 0.1 and 0.2 microM, respectively. This modified electrode was found to be very stable and reproducible when continuously used as detector for capillary electrophoresis for period of at least 4 weeks with no apparent loss of response.