Merck
  • Self-oligomerization and protein aggregation of alpha-synuclein in the presence of Coomassie Brilliant Blue.
Self-oligomerization and protein aggregation of alpha-synuclein in the presence of Coomassie Brilliant Blue.
European journal of biochemistry (2001-02-13)
D Lee, E K Lee, J H Lee, C S Chang, S R Paik
ABSTRACT
alpha-Synuclein has been implicated in various neurodegenerative disorders, including Parkinson's and Alzheimer's diseases, by its participation in abnormal protein depositions. As the protein has been suggested to play a significant role in the formation of the deposits which might be responsible for neurodegeneration, there is a strong demand to screen for alpha-synuclein-interactive small molecules. In this report, Coomassie Brilliant Blue (CBB) interaction of alpha-synuclein has been investigated with respect to induction of protein self-oligomerization in the presence of the chemical coupling reagent N-(ethoxycarbonyl)-2-ethoxy-1,2-dihydroquinoline. Both CBB-G and CBB-R, which differ by only two methyl groups, induced the self-oligomerization of alpha-synuclein in a biphasic manner with optimal dye concentrations of 250 microM and 150 microM, respectively. The protein aggregates of alpha-synuclein induced by the dyes in the absence of the coupling reagent were analysed by electron microscopy. Whereas CBB-G induced formation of protein aggregates with a worm-like structure, CBB-R induced clear fibrilization of alpha-synuclein on a background of granular structures. CBB-R interacted with alpha-synuclein approximately twice as effectively as CBB-G (dissociation constants 0.63 microM and 1.37 microM, respectively). These dye interactions were independent from the acidic C-terminus of alpha-synuclein, which was reminiscent of the Alphabeta25-35 interaction of alpha-synuclein. However, the metal-catalysed oxidative self-oligomerization of alpha-synuclein in the presence of Cu2+/H2O2, which was augmented synergistically by Alphabeta25-35, was not affected by the dyes. This indicates that the dye binding site is also distinctive from the Alphabeta25-35 interaction site on alpha-synuclein. These biochemically specific interactions between alpha-synuclein and the dyes indicate that alpha-synuclein-interactive small molecules could provide a tool with which to approach development of diagnostic, preventive, or therapeutic strategies for various alpha-synuclein-related neurodegenerative disorders.
Sigma-Aldrich
Brilliant Blue R Staining Solution, suitable for (for immunoelectrophoresis protein staining)
Sigma-Aldrich
Brilliant Blue R, pure
Sigma-Aldrich
Brilliant Blue R Concentrate, suitable for SDS-PAGE, methanol solution
Sigma-Aldrich
2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, ≥99%
Sigma-Aldrich
Brilliant Blue R, Dye content ~50 %, Technical grade
Sigma-Aldrich
Brilliant Blue R, 250, suitable for microscopy