Merck
  • Bis(mercaptoimidazolyl)(pyrazolyl)hydroborato complexes of zinc, cadmium, and cobalt: structural evidence for the enhanced tendency of zinc in biological systems to adopt tetrahedral M[S4] coordination.
Bis(mercaptoimidazolyl)(pyrazolyl)hydroborato complexes of zinc, cadmium, and cobalt: structural evidence for the enhanced tendency of zinc in biological systems to adopt tetrahedral M[S4] coordination.
Inorganic chemistry (2001-02-24)
C Kimblin, B M Bridgewater, D G Churchill, T Hascall, G Parkin
ABSTRACT
The bis(2-mercapto-1-methylimidazolyl)(pyrazolyl)hydroborato derivatives [pzBmMe]2Zn, [pzBmMe]2Co, and [pzBmMe]2Cd have been isolated and structurally characterized by X-ray diffraction. Despite their common [pzBmMe]2M composition, each of these complexes adopts a different structure. Thus, (i) the zinc complex exhibits a tetrahedral Zn[S4] structure in which only the sulfur donors coordinate to zinc, (ii) the cobalt complex exhibits a trigonal-bipyramidal Co[S3NH] structure in which one of the pyrazolyl groups and one of the B-H groups coordinate to cobalt, and (iii) the cadmium complex exhibits a six-coordinate Cd[S4H2] structure in which both B-H groups interact with the cadmium center. These comparisons emphasize that zinc has a greater preference for tetrahedral M[S4] coordination than does either cobalt or cadmium, an observation that is in accord with the prevalent role of zinc in the structural sites of enzymes.
Sigma-Aldrich
2-Mercaptoimidazole, 98%