Merck
  • [Simultaneous determination of fluoroacetamide and tetramine by gas chromatography].
[Simultaneous determination of fluoroacetamide and tetramine by gas chromatography].
Se pu = Chinese journal of chromatography (2003-01-25)
Qing Wu, Ming-shi Zhang, Zhao-rong Lan
ABSTRACT
A simple and fast gas chromatographic method was used for the simultaneous determination of fluoroacetamide and tetramine. The samples were extracted with acetone and the extracts were analyzed by gas chromatography. A megabore capillary column coated with FFAP and a nitrogen-phosphorous detector were used to obtain satisfactory selectivity and sensitivity. The detection limits and average recoveries were 5 ng and 69.1% for fluoroacetamide and 1 ng and 89.8% for tetramine.
Sigma-Aldrich
Fluoroacetamide