Merck
  • Liquid chromatographic determination of 4,4'-dinitrocarbanilide, the active component of the infertility agent nicarbazin, in chicken, duck, goose, and snake eggs.
Liquid chromatographic determination of 4,4'-dinitrocarbanilide, the active component of the infertility agent nicarbazin, in chicken, duck, goose, and snake eggs.
Journal of AOAC International (2004-02-26)
Thomas M Primus, Dennis J Kohler, Margaret A Goodall, Christi Yoder, Thomas Mathies, Lowell Miller, John J Johnston, Kurt Vercauteren
ABSTRACT
4,4'-Dinitrocarbanilide (DNC) was extracted from chicken, duck, goose, and snake eggs and isolated by reversed-phase liquid chromatography. DNC was detected by ultraviolet absorbance at 347 nm and quantitated by comparison with a calibration standard. Recoveries of DNC from fortified control chicken, duck, goose, and snake egg samples were determined for DNC levels of 0.16, 10, and 16 microg/g. The mean recoveries from chicken, duck, goose, and snake eggs were 92 +/- 4, 88 +/- 9, 87 +/- 7, and 95 +/- 6%, respectively. The method limits of detection for DNC in chicken, duck, goose, and snake eggs ranged from 0.015 to 0.035 microg/g. The reported method is much simpler than and equally efficient as previous methods developed for the determination of DNC residues in egg contents.
Sigma-Aldrich
1,3-Bis(4-nitrophenyl)urea, 97%
Supelco
Nicarbazin, VETRANAL®, analytical standard