Merck
  • A spectrophotometric micromethod for determining erythrocyte protoporphyrin-IX in whole blood or erythrocytes.
A spectrophotometric micromethod for determining erythrocyte protoporphyrin-IX in whole blood or erythrocytes.
Clinical chemistry and laboratory medicine (2005-04-22)
Guenther Kufner, Helmut Schlegel, Reinhard Jäger
ABSTRACT
An increased concentration of erythrocyte protoporphyrin-IX in whole blood or erythrocytes is a valuable diagnostic indicator for acquired porphyrias, e.g., iron deficiency anemia and lead poisoning, and for inherited porphyrias. We developed a spectrophotometric micromethod for determining erythrocyte protoporphyrin-IX. In this method, exhaustive release of erythrocyte porphyrins is achieved using hydroquinone and formic acid. The clean-up procedure for 50 microL of whole blood or erythrocytes covers three steps of liquid/liquid solvent partition: two partitions using diethyl and diisopropyl ether and HCl 2.5 mol/L, and one buffered step using ammonium formate. Determinations of erythrocyte protoporphyrin-IX are possible by: (a) absorption using three wavelengths, Rimington's constant and a millimolar absorptivity coefficient m epsilon(408.8)=294.3 L x mmol(-1) x cm(-1) according to With; and (b) 2nd derivative, which is linked to m epsilon(408.8). Determination of erythrocyte protoporphyrin-IX using a 2nd derivative algorithm showed better spectral resolution and higher sensitivity at a five-fold lower detection limit compared to absorption. Within-run precision of medium and high levels was found for absorption and for 2nd derivative with a coefficient of variation (CV) of 1.4-1.9% (n=10). Total precision evaluated was CV=2.5-8.3% (n=20). Levels of reference intervals could only be measured using the 2nd derivative (CV 2.9%). Linearity was proved to E=1.0. Recoveries of protoporphyrin-IX ranged from 95.3% to 103.0%. Method comparison was carried out using a fluorimetric reference method (Piomelli). Reference intervals for gender groups are discussed.
Product Description
Supelco
Diisopropyl ether, analytical standard
Sigma-Aldrich
Diisopropyl ether, anhydrous, 99%, contains either BHT or hydroquinone as stabilizer
Sigma-Aldrich
Diisopropyl ether, ReagentPlus®, 99%, contains either BHT or hydroquinone as stabilizer
Sigma-Aldrich
Diisopropyl ether, contains either BHT or hydroquinone as stabilizer, ACS reagent, ≥99.0%
Sigma-Aldrich
Diisopropyl ether, puriss. p.a., ≥98.5% (GC)