Merck
  • Direct cyclopropanation of 1-alkynylphosphonates by Cp2ZrCl2/2EtMgBr/2AlCl3 to afford cyclopropylmethylphosphonates.
Direct cyclopropanation of 1-alkynylphosphonates by Cp2ZrCl2/2EtMgBr/2AlCl3 to afford cyclopropylmethylphosphonates.
The Journal of organic chemistry (2006-01-18)
Abed Al Aziz Quntar, Morris Srebnik
ABSTRACT
[reaction: see text] The reagent system Cp2ZrCl2/2EtMgBr/2AlCl3 converts 1-alkynylphosphonates into cyclopropylmethylphosphonates 3 in good isolated yields. Ethers, chlorides, and other cyclopropyl groups are compatible with the reaction conditions. Deuterium labeling is consistent with the formation of stable cyclopropylmethylbimetallic phosphonates by ring contraction of the corresponding aluminacyclopentenylphosphonate. Temperature is crucial; apparently, the cyclopropylmethylbimetallic phosphonates are in equilibrium with the aluminacyclopentenylphosphonates. Low temperature favors the former. We surmise that the negative charges of the intermediate are stabilized by the phosphonate group. Thus, diphenylacetylene and 3-hexyne failed to give cyclopropyl products under the same reaction conditions.
Sigma-Aldrich
Zirconium(IV) chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Zirconium(IV) chloride, ≥99.9% trace metals basis
Sigma-Aldrich
Zirconium(IV) chloride, ≥99.5% trace metals basis