Merck
  • Supercritical fluid extraction (SFE) of 4(5)-methylimidazole (4-MeI) and 2-acetyl-4(5)-(1,2,3,4)-tetrahydroxybutyl-imidazole (THI) from ground-coffee with high-performance liquid chromatographic-electrospray mass spectrometric quantification (HPLC/ESI-MS).
Supercritical fluid extraction (SFE) of 4(5)-methylimidazole (4-MeI) and 2-acetyl-4(5)-(1,2,3,4)-tetrahydroxybutyl-imidazole (THI) from ground-coffee with high-performance liquid chromatographic-electrospray mass spectrometric quantification (HPLC/ESI-MS).
Food additives and contaminants (2006-09-20)
L Lojková, B Klejdus, J Moravcová, V Kubán
ABSTRACT
Two polar analytes, 4(5)-methylimidazole (4-MeI) and 2-acetyl-4(5)-(1,2,3,4)-tetrahydroxybutyl-imidazole (THI), were extracted with supercritical carbon dioxide (CO2) modified with aqueous methanol. The method was applied to a roasted coffee powder with good recovery rates. Method efficiency was compared with that of solid-phase extraction using SCX Disc cartridges and validated for spiked solid matrix. The analytes were determined using isocratic liquid chromatography-mass spectrometry (LC/MS) on an Atlantis HILIC Silica column (150 x 2.1 mm, 3 microm) with 80% methanol and 20% 0.01 mol l-1 ammonium formate as the mobile phase. The limit of quantification was around 1.5 pg for 4-MeI and 2.0 pg for THI. The linearity of the calibration curves was satisfactory as indicated by correlation coefficients of >0.999. The coefficient of variation for the intra-day and inter-day precisions was <4% (n = 6). Accuracy was in the range 98-101%; recovery rates were > or = 98 and > or = 99% for THI and 4-MeI, respectively. Several samples of Arabica coffee from various locations and commercially available 'off-the-shelf' coffee products (Arabica/Robusta mixtures) were analysed to test the method.
Sigma-Aldrich
THI
Sigma-Aldrich
4(5)-Methylimidazole, 98%