Merck
  • An efficient synthesis of enantiomerically pure (R)-pipecolic acid, (S)-proline, and their N-alkylated derivatives.
An efficient synthesis of enantiomerically pure (R)-pipecolic acid, (S)-proline, and their N-alkylated derivatives.
The Journal of organic chemistry (2007-01-30)
Antoine Fadel, Nabil Lahrache
ABSTRACT
Enantiomerically pure (R)-(+)-pipecolic acid was synthesized in four steps and 42% overall yield starting from dihydropyran and (R)-alpha-methylbenzylamine. A general short strategy is also described for preparing (S)-proline (47.5% overall yield) and derivatives.
Product Description
Supelco
(R)-(+)-α-Methylbenzylamine, derivatization grade (chiral), LiChropur, ≥99.0%
Sigma-Aldrich
(R)-(+)-α-Methylbenzylamine, purum, ≥98.0% (sum of enantiomers, GC)
Sigma-Aldrich
α-Methylbenzylamine, 99%
Sigma-Aldrich
(R)-(+)-α-Methylbenzylamine, ChiPros®, produced by BASF, ≥99.0%
Sigma-Aldrich
(R)-(+)-α-Methylbenzylamine, 98%
Sigma-Aldrich
(S)-(−)-α-Methylbenzylamine, 98%