Merck
  • SanM catalyzes the formation of 4-pyridyl-2-oxo-4-hydroxyisovalerate in nikkomycin biosynthesis by interacting with SanN.
SanM catalyzes the formation of 4-pyridyl-2-oxo-4-hydroxyisovalerate in nikkomycin biosynthesis by interacting with SanN.
Biochemical and biophysical research communications (2007-07-31)
Hongbo Ling, Guojun Wang, Yuqing Tian, Gang Liu, Huarong Tan
ABSTRACT
Nikkomycins are peptidyl nucleoside antibiotics with potent activities against phytopathogenic and human pathogenic fungi. The sanM and sanN genes are required for the nikkomycin biosynthesis of Streptomyces ansochromogenes. In the present study, interaction between SanM and SanN was identified by yeast two-hybrid and co-immunoprecipitation assays. Moreover, SanM and SanN were heterologously expressed and purified. Further biochemical assay demonstrated that the SanM-SanN interaction is essential for SanM aldolase activity but not for SanN dehydrogenase activity. SanM converts piconaldehyde and 2-oxobutyrate to 4-pyridyl-2-oxo-4-hydroxyisovalerate in nikkomycin biosynthesis by interacting with SanN. Steady state kinetics analysis revealed that K(m) and k(cat)/K(m) of SanM are 123.2 microM and 11.4 mM(-1)s(-1) for picolinaldehyde, while 335.6 microM and 4.0 mM(-1)s(-1) for 2-oxobutyrate, respectively. However, SanN as a dehydrogenase is independent of SanM.
Sigma-Aldrich
Sodium 2-oxobutyrate, powder
Sigma-Aldrich
2-Oxobutyric acid, ≥95%, FG
Sigma-Aldrich
2-Ketobutyric acid, 97%