Merck
  • Unexpected C-H activation of Ru(II)-dithiomaltol complexes upon oxidation.
Unexpected C-H activation of Ru(II)-dithiomaltol complexes upon oxidation.
Inorganic chemistry (2008-02-09)
Malin Backlund, Joseph Ziller, Patrick J Farmer
ABSTRACT
Thione-substituted derivatives of maltol are of interest in several applications of metal-based drugs. In order to investigate the effect of the oxygenation on such thione chelates, Ru complexes of 3-hydroxy-2-methyl-4-thiopyrone (thiomaltol or Htma) and 3-hydroxy-2-methyl-4H-thiopyran-4-thione (dithiomaltol or Httma), [Ru(bpy)2(tma)](+), 1, and [Ru(bpy) 2(ttma)] (+), 2, were synthesized as diamagnetic PF6(-) salts. Peroxidation of 2 unexpectedly generated products of C-H activation at its pendant methyl group; an air-stable aldehyde [Ru(bpy)2(ttma-aldehyde)](+), 4, was the major product. In addition, an intermediate oxidation product [Ru(bpy) 2(ttma-alcohol)](PF6), 3, was characterized. Both 3 and 4 are also formed by reaction of 2 with outersphere oxidants (e.g., Na2IrCl6) and by bulk electrolysis under anaerobic conditions. Similar oxidations of the analogous [Ru(bpy)2(ettma)](+), 2' , complex (3-hydroxy-2-ethyl-4H-thiopyran-4-thione; ethyl dithiomaltol or Hettma) formed the corresponding ketone, [Ru(bpy)2(ettma-ketone)](PF6), 4', by oxidation at the same position adjacent to the conjugated ring. The structures of the aldehyde 4 and starting materials 1 and 2 have been confirmed by X-ray crystallography, and all complexes have been characterized by UV-vis, (1)H NMR, and IR spectroscopies. Initial mechanistic investigations are discussed.
Sigma-Aldrich
3-Hydroxy-2-methyl-4-pyrone, 99%
Sigma-Aldrich
Maltol, ≥99.0%, FCC, FG
Sigma-Aldrich
Maltol, natural, FG