Merck
  • Novel thermochromism relating to supramolecular cuprophilic interaction: design, synthesis, and luminescence of copper(I) pyrazolate trimer and polymer.
Novel thermochromism relating to supramolecular cuprophilic interaction: design, synthesis, and luminescence of copper(I) pyrazolate trimer and polymer.
Inorganic chemistry (2008-04-03)
Jing-Xiang Zhang, Jun He, Ye-Gao Yin, Mei-Hong Hu, Dan Li, Xiao-Chun Huang
ABSTRACT
Solvothermal reactions of 4-(pyrid-4'-yl)-3,5-dimethylpyrazole (HPpz) with CuBr in two mixed solvents, NH3.H2O/EtOH and NH3.H2O/MeCN, afforded respectively a copper(I) trimer, [Cu(Ppz)]3(1), and a polymer, {[Cu(Ppz)]3[CuCN] 3} (2), both containing the [Cu(Ppz)]3 entity as a building block. The products were found to be photoluminescent and, more interestingly, when cooled from room temperature to 10 K, they showed a blue shift followed by a red shift (hereafter shortened to a red-after-blue shift) of emission.
Sigma-Aldrich
Copper(II) bromide, 99%
Sigma-Aldrich
Copper(II) bromide, 99.999% trace metals basis