Merck
  • [Determination of five primary chemical constituents in Cassia angustifolia by HPLC].
[Determination of five primary chemical constituents in Cassia angustifolia by HPLC].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2008-06-07)
Qiu-Ping Wu, Zhu-Ju Wang, Li-Ying Tang, Mei-Hong Fu, Yan He, Jing Fang, Qian-Feng Gong
ABSTRACT
To establish a method for determining the content of primary chemical constituents in the leaves of Cassia angustifolia. The HPLC with Diamonsil C18 (4.6 mm x 250 mm, 5 microm) column was used , acetonitrile-1% acetic acid (10:90-15: 85-18: 82-20: 80-25: 75) in a gradient manner was used as a mobile phase, with flow rate of 1 mL x min(-1), column temperature at 40 degrees C and detection wavelength at 270 nm. The results showed that 5 effective components all separated well and showed good linearity. The method was proved to be rapid, sensitive, accurate, credible and repeatable. It can be applied to quality control of Folium Sennae.
Product Description
Supelco
Sennoside A, analytical standard
Sennoside B, primary reference standard