Merck
  • 100,000-fold concentration of anions in capillary zone electrophoresis using electroosmotic flow controlled counterflow isotachophoretic stacking under field amplified conditions.
100,000-fold concentration of anions in capillary zone electrophoresis using electroosmotic flow controlled counterflow isotachophoretic stacking under field amplified conditions.
Analytical chemistry (2008-07-17)
Michael C Breadmore, Joselito P Quirino
ABSTRACT
An electroosmotic flow (EOF) controlled counterflow isotachophoretic stacking boundary (cf-ITPSB) system under field amplified conditions has been examined as a way to improve the sensitivity of anions separated by capillary zone electrophoresis. The system comprised a high concentration of a high-mobility leading ion (100 mM chloride) and a low concentration of low-mobility terminating ion (1-3 mM MES or CHES) added to the sample in an unmodified fused-silica capillary at pH 8.05, buffered with Tris. Computer simulation studies using the software GENTRANS showed an increase in sensitivity of at least 10-fold over the previous cf-ITPSB system for simple inorganic ions, nitrite and nitrate. The simulations also suggested that the cf-ITPSB became stationary within the capillary and that its stationary position was not adversely affected by the concentration of MES. This was in contrast to experimental results that showed a slow and continual movement of the cf-ITPSB. This was more pronounced at lower concentrations of terminator (i.e., <3 mM) and resulted in a loss of resolution due to the cf-ITPSB being closer to the detector upon separation. This discrepancy was attributed to the change in pH across the capillary due to electrolysis and low buffering capacity in the sample, a phenomenon that cannot be simulated by the GENTRANS software. Replacement of MES with CHES as a lower mobility ion with increased buffer capacity failed to reduce the movement of the cf-ITPSB but did provide a further 3-fold improvement in sensitivity. The potential of this approach for sensitivity enhancement was demonstrated for the co-EOF separation of a mixture of six inorganic and small organic ions, with detection limits at the single-figure nanogram per liter level. These detection limits are 100,000 times better than can be achieved by normal hydrodynamic injection (ions prepared in water) and 250 times better than has been achieved by other online preconcentration approaches. The application of the EOF-controlled cf-ITPSB with counter-EOF separation of two pharmaceutical pollutants, naproxen and diflunisal, was also demonstrated with an improvement in sensitivity of 1000 giving detection limits of 350 ng/L in sewage treatment wastewater without any offline pretreatment.
Product Description
SAFC
MES sodium salt
Sigma-Aldrich
MES solution, BioUltra, Molecular Biology, 0.5 M in H2O
Sigma-Aldrich
CHES, BioUltra, ≥99.5% (T)
Sigma-Aldrich
MES sodium salt, BioReagent, ≥99% (titration), crystalline
Sigma-Aldrich
MES sodium salt, ≥99% (titration)
Sigma-Aldrich
MES sodium salt, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
CHES, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
CHES, ≥99.0% (titration)
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
MES hydrate, BioPerformance Certified, suitable for cell culture, ≥99.5%
Sigma-Aldrich
MES hydrate, ≥99.5% (titration), pH 2.5-4.0 (0.5 M in H2O), BioXtra
Sigma-Aldrich
MES hydrate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
MES hydrate, ≥99.5% (titration)