Merck
  • Multi-channel open tubular traps for headspace sampling, gas chromatographic fraction collection and olfactory assessment of milk volatiles.
Multi-channel open tubular traps for headspace sampling, gas chromatographic fraction collection and olfactory assessment of milk volatiles.
Journal of chromatography. A (2008-10-22)
Yvette Naudé, Marleen van Aardt, Egmont Richard Rohwer
ABSTRACT
A headspace sampling method is described for concentrating milk volatiles onto a multi-channel open tubular silicone rubber trap (MCT) for thermal desorption into a GC-FID. Sections of the chromatographic profile, single peaks or combinations of compounds are recaptured with secondary MCTs during a subsequent run. The recaptured aroma is released in a controlled manner by heating the MCT in a portable heating device. An aroma release window of several minutes allows up to six people the opportunity to sniff each aroma fraction more than once. Olfactory results suggest that a synergistic combination of 2-heptanone and 2-nonanone could be responsible for a pungent cheese, sour milk-like aroma. MCTs containing single components or fractions can be desorbed into a GC-MS for compound identification.
Sigma-Aldrich
2-Heptanone, 99%
Sigma-Aldrich
2-Nonanone, ≥99%
Supelco
2-Heptanone, analytical standard
Sigma-Aldrich
2-Nonanone, natural, ≥97%, FCC, FG
Sigma-Aldrich
2-Heptanone, ≥98%, FCC, FG
Sigma-Aldrich
2-Nonanone, ≥99%, FCC, FG