Merck
  • Desymmetrization of cyclic anhydrides mediated by cinchona alkaloids: synthesis and olfactory properties of new fragrances based on (R)- and (S)-2-ethylhexanol.
Desymmetrization of cyclic anhydrides mediated by cinchona alkaloids: synthesis and olfactory properties of new fragrances based on (R)- and (S)-2-ethylhexanol.
Chirality (2009-01-24)
Blazenka Cisko-Anić, Zdenko Hamersak
ABSTRACT
A series of enantiomerically pure new fragrances, derived from 2-ethylhexanol, have been prepared and their olfactory properties evaluated. The key step of the synthesis is cinchona-alkaloid-catalyzed desymmetrization of cyclic meso-anhydrides with (R)- and (S)-2-ethylhexanol and proceeded in good to excellent diastereoselectivities (92:8-98:2 dr). Enantiomerically pure alcohols were prepared by lipase-catalyzed kinetic resolution of 2-ethylhexanol using vinyl laurate as acyl donor.
Product Description
Supelco
2-Ethyl-1-hexanol, analytical standard
Sigma-Aldrich
2-Ethyl-1-hexanol, ≥99%, FG
Sigma-Aldrich
2-Ethyl-1-hexanol, puriss., ≥99.0% (GC)
Sigma-Aldrich
2-Ethyl-1-hexanol, ≥99.6%
Sigma-Aldrich
Titanium(IV) 2-ethylhexyloxide, 95%