Merck
  • Aliphatic C-H bond activation initiated by a (mu-eta2:eta2-peroxo)dicopper(II) complex in comparison with cumylperoxyl radical.
Aliphatic C-H bond activation initiated by a (mu-eta2:eta2-peroxo)dicopper(II) complex in comparison with cumylperoxyl radical.
Journal of the American Chemical Society (2009-06-18)
Takahiro Matsumoto, Kei Ohkubo, Kaoru Honda, Akiko Yazawa, Hideki Furutachi, Shuhei Fujinami, Shunichi Fukuzumi, Masatatsu Suzuki
ABSTRACT
A (mu-eta(2):eta(2)-peroxo)dicopper(II) complex, [Cu(2)(H-L)(O(2))](2+) (1-O(2)), supported by the dinucleating ligand 1,3-bis[bis(6-methyl-2-pyridylmethyl)aminomethyl]benzene (H-L) is capable of initiating C-H bond activation of a variety of external aliphatic substrates (SH(n)): 10-methyl-9,10-dihydroacridine (AcrH(2)), 1,4-cyclohexadiene (1,4-CHD), 9,10-dihydroanthracene (9,10-DHA), fluorene, tetralin, toluene, and tetrahydrofuran (THF), which have C-H bond dissociation energies (BDEs) ranging from approximately 75 kcal mol(-1) for 1,4-CHD to approximately 92 kcal mol(-1) for THF. Oxidation of SH(n) afforded a variety of oxidation products, such as dehydrogenation products (SH((n-2))), hydroxylated and further-oxidized products (SH((n-1))OH and SH((n-2))=O), dimers formed by coupling between substrates (H((n-1))S-SH((n-1))) and between substrate and H-L (H-L-SH((n-1))). Kinetic studies of the oxidation of the substrates initiated by 1-O(2) in acetone at -70 degrees C revealed that there is a linear correlation between the logarithms of the rate constants for oxidation of the C-H bonds of the substrates and their BDEs, except for THF. The combination of this correlation and the relatively large deuterium kinetic isotope effects (KIEs), k(2)(H)/k(2)(D) (13 for 9,10-DHA, approximately > 29 for toluene, and approximately 34 for THF at -70 degrees C and approximately 9 for AcrH(2) at -94 degrees C) indicates that H-atom transfer (HAT) from SH(n) (SD(n)) is the rate-determining step. Kinetic studies of the oxidation of SH(n) by cumylperoxyl radical showed a correlation similar to that observed for 1-O(2), indicating that the reactivity of 1-O(2) is similar to that of cumylperoxyl radical. Thus, 1-O(2) is capable of initiating a wide range of oxidation reactions, including oxidation of aliphatic C-H bonds having BDEs from approximately 75 to approximately 92 kcal mol(-1), hydroxylation of the m-xylyl linker of H-L, and epoxidation of styrene (Matsumoto, T.; et al. J. Am. Chem. Soc. 2006, 128, 3874).
Sigma-Aldrich
1,4-Cyclohexadiene, 97%
Sigma-Aldrich
1,4-Cyclohexadiene, purum, ≥97.0% (GC)
Sigma-Aldrich
1,2,3,4-Tetrahydronaphthalene, ReagentPlus®, 99%
Sigma-Aldrich
1,2,3,4-Tetrahydronaphthalene, reagent grade, ≥97%
Sigma-Aldrich
1,2,3,4-Tetrahydronaphthalene, anhydrous, 99%
Supelco
1,2,3,4-Tetrahydronaphthalene, analytical standard