Merck
  • Chitosan derivative nanocarrier: safety evaluation,antibacterial property and ascorbyl palmitate encapsulation.
Chitosan derivative nanocarrier: safety evaluation,antibacterial property and ascorbyl palmitate encapsulation.
Journal of microencapsulation (2009-07-02)
Mayura Wittayasuporn, Sirirat Rengpipat, Tanapat Palaga, Pravit Asawanonda, Nattaporn Anumansirikul, Supasorn P Wanichwecharungruang
ABSTRACT
A chitosan derivative, methyl ether-terminated poly(ethylene oxide)-4-methoxycinnamolyphthaloylchitosan (PCPLC) was prepared, characterized and self-assembled into nanoparticles. Encapsulation of ascorbyl palmitate (AP) into PCPLC gave 6890.98 nm particles with encapsulation efficiency of 84% at 56% drug loading. The encapsulated AP showed significant improved stability as examined by 1H NMR spectroscopy.The obtained particles displayed no short-term cytotoxicity against the human skin melanoma A-375 cell line using the MTT assay and no short-term skin irritation on human volunteers using a single topical application as patch and photopatch tests. In addition, aqueous suspension of PCPLC nanoparticles successfully inhibited the growth of Escherichia coli ATCC 25922 and Staphylococcus aureus ATCC 25923.
Sigma-Aldrich
6-O-Palmitoyl-L-ascorbic acid, BioXtra, ≥99.0% (RT)
Sigma-Aldrich
Ascorbic acid 6-palmitate, meets USP testing specifications