Merck
  • Aromatic hydroxylation of indan by o-xylene-degrading Rhodococcus sp. strain DK17.
Aromatic hydroxylation of indan by o-xylene-degrading Rhodococcus sp. strain DK17.
Applied and environmental microbiology (2009-11-03)
Dockyu Kim, Choong Hwan Lee, Jung Nam Choi, Ki Young Choi, Gerben J Zylstra, Eungbin Kim
ABSTRACT
The metabolically versatile Rhodococcus sp. strain DK17 utilizes indan as a growth substrate via the o-xylene pathway. Metabolite and reverse transcription-PCR analyses indicate that o-xylene dioxygenase hydroxylates indan at the 4,5 position of the aromatic moiety to form cis-indan-4,5-dihydrodiol, which is dehydrogenated to 4,5-indandiol by a dehydrogenase. 4,5-indandiol undergoes ring cleavage by a meta-cleavage dioxygenase.
Sigma-Aldrich
Indan, 95%
Sigma-Aldrich
Xylenes, ACS reagent, ≥98.5% xylenes + ethylbenzene basis
Sigma-Aldrich
Xylenes, reagent grade
Sigma-Aldrich
Xylenes, histological grade
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
o-Xylene, analytical standard
Supelco
Indan, analytical standard