Merck
  • Oriented synthesis and in vitro anticancer activity of biquinazoline-2,2'-diones.
Oriented synthesis and in vitro anticancer activity of biquinazoline-2,2'-diones.
Journal of combinatorial chemistry (2009-12-24)
Guolan Dou, Daqing Shi, Yonghai Li
ABSTRACT
The synthesis of a series of biquinazoline-2,2'-diones starting from o-nitrobenzaldehydes, anilines, and triphosgene is presented. This general approach features a novel and easy way for access to the target products. The mechanistic course of the reaction suggests the involvement of reduction, coupling, and cyclization by one-pot. These compounds were also investigated in vitro for anticancer activity, and some were found to have good anticancer activity.
Sigma-Aldrich
Triphosgene, reagent grade, 98%
Sigma-Aldrich
2-Nitrobenzaldehyde, 98%
Sigma-Aldrich
Bis(trichloromethyl) carbonate, purum, ≥99.0% (AT)