Merck
  • Transport and degradation of pesticides in a biopurification system under variable flux, Part I: a microcosm study.
Transport and degradation of pesticides in a biopurification system under variable flux, Part I: a microcosm study.
Environmental pollution (Barking, Essex : 1987) (2010-08-11)
Tineke De Wilde, Pieter Spanoghe, Jaak Ryckeboer, Peter Jaeken, Dirk Springael
ABSTRACT
The efficiency of a biopurification system, developed to treat pesticide contaminated water, is to a large extent determined by the chemical and hydraulic load. Insight into the behaviour of pesticides under different fluxes is necessary. The behaviour of metalaxyl, bentazone, linuron, isoproturon and metamitron was studied under three different fluxes with or without the presence of pesticide-primed soil in column experiments. Due to the time-dependent sorption process, retention of the pesticides with intermediate mobility was significantly influenced by the flux. The higher the flux, the slower pesticides will be sorbed, which resulted in a lower retention. Degradation of the intermediate mobile pesticides was also submissive to variations in flux. An increase in flux, led to a decrease in retention, which in turn decreased the opportunity time for biodegradation. Finally, the presence of pesticide-primed soil was only beneficial for the degradation of metalaxyl.
Product Description
Supelco
Linuron, PESTANAL®, analytical standard
Supelco
Metamitron, PESTANAL®, analytical standard
Supelco
Bentazon, PESTANAL®, analytical standard
Supelco
Metalaxyl, PESTANAL®, analytical standard