Merck
  • Elastin peptides signaling relies on neuraminidase-1-dependent lactosylceramide generation.
Elastin peptides signaling relies on neuraminidase-1-dependent lactosylceramide generation.
PloS one (2010-11-26)
Anthony Rusciani, Laurent Duca, Hervé Sartelet, Aurore Chatron-Colliet, Hélène Bobichon, Dominique Ploton, Richard Le Naour, Sébastien Blaise, Laurent Martiny, Laurent Debelle
ABSTRACT
The sialidase activity of neuraminidase-1 (Neu-1) is responsible for ERK 1/2 pathway activation following binding of elastin peptide on the elastin receptor complex. In this work, we demonstrate that the receptor and lipid rafts colocalize at the plasma membrane. We also show that the disruption of these microdomains as well as their depletion in glycolipids blocks the receptor signaling. Following elastin peptide treatment, the cellular GM(3) level decreases while lactosylceramide (LacCer) content increases consistently with a GM(3)/LacCer conversion. The use of lactose or Neu-1 siRNA blocks this process suggesting that the elastin receptor complex is responsible for this lipid conversion. Flow cytometry analysis confirms this elastin peptide-driven LacCer generation. Further, the use of a monoclonal anti-GM(3) blocking antibody shows that GM(3) is required for signaling. In conclusion, our data strongly suggest that Neu-1-dependent GM(3)/LacCer conversion is the key event leading to signaling by the elastin receptor complex. As a consequence, we propose that LacCer is an early messenger for this receptor.
Sigma-Aldrich
Anti-NEU1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution