Merck
  • Simultaneous hyperaccumulation of multiple heavy metals by Helianthus annuus grown in a contaminated sandy-loam soil.
Simultaneous hyperaccumulation of multiple heavy metals by Helianthus annuus grown in a contaminated sandy-loam soil.
International journal of phytoremediation (2010-12-21)
Teresa Cutright, Nagaraju Gunda, Firat Kurt
ABSTRACT
Phytoremediation is a promising means for the treatment of contamination arising from heavy metal spills. Although several species have been identified as hyperaccumulators, most of the studies were performed with only one heavy metal. Experiments were conducted with two cultivars of H. annuus exposed to different combinations of metal contamination (30 mg/kg Cd, Cr, Ni, As, and/or Fe). Cultivar efficiency was based on total metal uptake, as well as translocation and selectivity of each metal. The results for each cultivar were also compared after 0.1 g/kg or 0.3 g/kg EDTA was added to enhance metal bioavailability. The key finding was that H. annuus achieved hyperaccumulator status for multiple metals simultaneously: Cd, Cr, and As.
Sigma-Aldrich
Chromium(II) chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Chromium(II) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Chromium(II) chloride, 95%
Sigma-Aldrich
Cadmium sulfate 8/3-hydrate, puriss. p.a., ACS reagent, ≥99.0% (calc. based on CdSO4 · 8/3 H2O, KT)
Sigma-Aldrich
Cadmium sulfate, ACS reagent, ≥99.0%