Merck
  • Enantiospecific (S)-(+)-linalool formation from beta-myrcene by linalool dehydratase-isomerase.
Enantiospecific (S)-(+)-linalool formation from beta-myrcene by linalool dehydratase-isomerase.
Zeitschrift fur Naturforschung. C, Journal of biosciences (2011-09-29)
Frauke Lüddeke, Jens Harder
ABSTRACT
The linalool dehydratase-isomerase from Castellaniella defragrans strain 65Phen catalyzes in the thermodynamically unfavourable direction the hydration of betamyrcene to linalool and further the isomerization to geraniol, the initial steps in anaerobic beta-myrcene biodegradation. We have now investigated the stereochemistry of this reaction. (S)-(+)-Linalool is formed with an enantiomeric excess of at least 95.4%. (R)-(-)-Linalool was not detected. This indicates an introduction of the hydroxy group on the si-face of beta-myrcene.
Sigma-Aldrich
Myrcene, technical grade
Sigma-Aldrich
Myrcene, stabilized, FCC, FG
Supelco
Myrcene, analytical standard