Merck
  • Organ specificity of the bladder carcinogen 4-aminobiphenyl in inducing DNA damage and mutation in mice.
Organ specificity of the bladder carcinogen 4-aminobiphenyl in inducing DNA damage and mutation in mice.
Cancer prevention research (Philadelphia, Pa.) (2011-11-17)
Jae-In Yoon, Sang-In Kim, Stella Tommasi, Ahmad Besaratinia
ABSTRACT
Aromatic amines are a widespread class of environmental contaminants present in various occupational settings and tobacco smoke. Exposure to aromatic amines is a major risk factor for bladder cancer development. The etiologic involvement of aromatic amines in the genesis of bladder cancer is attributable to their ability to form DNA adducts, which upon eluding repair and causing mispairing during replication, may initiate mutagenesis. We have investigated the induction of DNA adducts in relation to mutagenesis in bladder and various nontarget organs of transgenic Big Blue mice treated weekly (i.p.) with a representative aromatic amine compound, 4-aminobiphenyl (4-ABP), for six weeks, followed by a six-week recovery period. We show an organ-specificity of 4-ABP in inducing repair-resistant DNA adducts in bladder, kidney, and liver of carcinogen-treated animals, which accords with the bioactivation pathway of this chemical in the respective organs. In confirmation, we show a predominant and sustained mutagenic effect of 4-ABP in bladder, and much weaker but significant mutagenicity of 4-ABP in the kidney and liver of carcinogen-treated mice, as reflected by the elevation of background cII mutant frequency in the respective organs. The spectrum of mutations produced in bladder of 4-ABP-treated mice matches the known mutagenic properties of 4-ABP-DNA adducts, as verified by the preponderance of induced mutations occurring at G:C base pairs (82.9%), with the vast majority being G:C→T:A transversions (47.1%). Our data support a possible etiologic role of 4-ABP in bladder carcinogenesis and provide a mechanistic view on how DNA adduct-driven mutagenesis, specifically targeted to bladder urothelium, may account for organ-specific tumorigenicity of this chemical.
Product Description
Supelco
4-Aminobiphenyl, analytical standard
Sigma-Aldrich
4-Aminobiphenyl, ≥98%