Merck
  • Vectorial release of matrix metalloproteinases (MMPs) from porcine RPE-choroid explants following selective retina therapy (SRT): towards slowing the macular ageing process.
Vectorial release of matrix metalloproteinases (MMPs) from porcine RPE-choroid explants following selective retina therapy (SRT): towards slowing the macular ageing process.
Experimental eye research (2012-03-06)
F Treumer, A Klettner, J Baltz, A A Hussain, Y Miura, R Brinkmann, J Roider, J Hillenkamp
ABSTRACT
The purpose of this study was to investigate release of matrix metalloproteinases (MMP) 2 and 9 during retinal pigment epithelium (RPE) wound healing after Selective Retina Therapy (SRT) with laser energy levels below and above the threshold of RPE cell death. Following exposure to SRT using a prototype pulsed Nd:YLF laser with energies of 80-180 mJ/cm(2) fresh porcine RPE-monolayers with Bruch's membrane and choroid were cultured in modified Ussing chambers which separate the apical (RPE-facing) and basal (choroid facing) sides of the RPE monolayer. Threshold energy for RPE cell death and wound healing were determined with calcein-AM viability test. Inactive and active forms of MMP 2 and 9 were quantified within tissue samples and in the culture medium of the apical and basal compartments of the Ussing chamber using gelatine zymography. Laser energies of 160-180 mJ/cm(2) resulted in cell death within 1 h while 120-140 mJ/cm(2) resulted in delayed death of exposed RPE cells. All cells survived 80 and 100 mJ/cm(2). Laser spots healed within 6 days after SRT accompanied by a transient vectorial increase of MMPs. SRT with 180 mJ/cm(2) increased active MMP 2 by 1.9 (p < 0.05) and 1.6 (p < 0.05) fold in tissue and basal compartments, respectively, without alterations in the apical compartment. Pro-MMP 2 levels were also significantly increased in all compartments (p < 0.05). Release of MMP 9 was not altered. Laser energy below the threshold of RPE cell death did not alter the release of MMP 2 or 9. The findings suggest that the release of active MMP 2 on the basal side of the RPE during wound healing following SRT may address age-related pathological changes of Bruch's membrane with a potential to slow degenerative macular ageing processes before irreversible functional loss has occurred.
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution