Merck
  • Reproducible vapor-time profiles using solid-phase microextraction with an externally sampled internal standard.
Reproducible vapor-time profiles using solid-phase microextraction with an externally sampled internal standard.
Journal of chromatography. A (2012-05-29)
William MacCrehan, Stephanie Moore, Michele Schantz
ABSTRACT
Determination of the dynamic nature of vapor/odor release has application in a wide variety of systems. This study applies automated solid-phase microextraction (SPME) utilizing an externally sampled internal standard (ESIS) to determine the vapor-time profile of odor delivery devices for three classes of explosive compounds. The profiles of delivery systems for target odorants 2,4-dinitrotoluene (2,4-DNT), 2-ethyl-1-hexanol (2-EH), and triacetone triperoxide (TATP) as canine training aids were compared over a period of 70 h. Strategies for evaluating the vapor-time profile of components with widely differing volatility are considered. An approach to quantifying the vapor concentration is described. The differences in the vapor-time profiles are examined and suggestions for selecting the best representative odor delivery technique are outlined.
Sigma-Aldrich
2,4-Dinitrotoluene, 97%
Supelco
2-Ethyl-1-hexanol, analytical standard
Sigma-Aldrich
2-Ethyl-1-hexanol, ≥99%, FG
Sigma-Aldrich
2-Ethyl-1-hexanol, puriss., ≥99.0% (GC)
Sigma-Aldrich
2-Ethyl-1-hexanol, ≥99.6%
Sigma-Aldrich
Titanium(IV) 2-ethylhexyloxide, 95%