Merck
  • Decylglucoside-based microemulsions for cutaneous localization of lycopene and ascorbic acid.
Decylglucoside-based microemulsions for cutaneous localization of lycopene and ascorbic acid.
International journal of pharmaceutics (2012-06-14)
Dominique Pepe, Jessica Phelps, Kevin Lewis, Jared Dujack, Katrina Scarlett, Sumaya Jahan, Emilee Bonnier, Tatjana Milic-Pasetto, Martha A Hass, Luciana B Lopes
ABSTRACT
Cutaneous delivery of combinations of antioxidants offers the possibility of enhanced protection against UV-radiation. In this study, we investigated the potential of sugar-based microemulsions containing monoglycerides to promote simultaneous cutaneous delivery of lycopene and ascorbic acid, and increase tissue antioxidant activity. Lycopene and ascorbic acid were incorporated (0.04% and 0.2% (w/w), respectively) in decylglucoside-based microemulsions containing isopropyl myristate mixed with monocaprylin (ME-MC), monolaurin (ME-ML) or monoolein (ME-MO) as oil phase. The microemulsions increased lycopene delivery into porcine ear skin by 3.3- to 8-fold compared to a drug solution. The effect of microemulsions on ascorbic acid cutaneous delivery was more modest (1.5-3-fold), and associated with an approximately 2-fold increase in transdermal delivery. According to their penetration-enhancing ability, the microemulsions were ranked ME-MC>ME-MO>ME-ML. This superiority of ME-MC coincided with a stronger effect in decreasing skin electrical resistance. After 18 h of treatment, the viability of bioengineered skin treated with ME-MC was 2.2-times higher compared to Triton-X100 (moderate irritant), demonstrating that ME-MC is less cytotoxic. Skin treatment with ME-MC containing both antioxidants increased the tissue antioxidant activity by 10.2-fold, but no synergism between the antioxidants was observed.
Product Description
Glycerol mono-oleate, European Pharmacopoeia (EP) Reference Standard
Supelco
Isopropyl myristate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Isopropyl myristate, 98%
Sigma-Aldrich
Isopropyl myristate, ≥90% (GC)
Sigma-Aldrich
Isopropyl myristate, ≥98%