Merck
  • Drug delivery systems for the topical treatment of cutaneous leishmaniasis.
Drug delivery systems for the topical treatment of cutaneous leishmaniasis.
Expert opinion on drug delivery (2012-06-26)
Guilherme Carneiro, Marta Gontijo Aguiar, Ana Paula Fernandes, Lucas Antônio Miranda Ferreira
ABSTRACT
The parenteral administration of pentavalent antimonials for the treatment of all forms of leishmaniasis, including cutaneous leishamniasis (CL), has several limitations. Therapy is long, requiring repeated doses and the adverse reactions are frequent. Topical treatment is an attractive alternative for CL, offering significant advantages over systemic therapy: fewer adverse effects, ease of administration, and lower costs. This review covers, from 1984 to the present, the progress achieved for the development of topical treatment for CL, using different drugs such as paromomycin (PA), imiquimod, amphotericin B (AmB), miltefosine, and buparvaquone. PA is the most commonly studied drug, followed by AmB and Imiquimod. These drugs were incorporated in conventional dosage forms or loaded in lipid nanocarries, which have been used mainly for improved skin delivery and antileishmanial activity. Developing an effective topical treatment for CL using these antileishmanial drugs still remains a great challenge. Insights into the most promising delivery strategies to improve treatment of CL with PA and AmB using conventional dosage forms, lipid nanocarriers, and combined therapy are presented and discussed. The results obtained with combined therapy and alternative delivery systems are promising perspectives for improving topical treatment of CL.
Sigma-Aldrich
Paromomycin sulfate salt, suitable for plant cell culture, BioReagent
Sigma-Aldrich
Paromomycin sulfate salt, powder, BioReagent, suitable for cell culture, potency: ≥675 μg per mg
Sigma-Aldrich
Paromomycin sulfate salt, ≥98% (TLC)
Supelco
Buparvaquone, VETRANAL®, analytical standard