Merck
Absorption spectra of PTCDI: a combined UV-Vis and TD-DFT study.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-08-14)
Mircea Oltean, Adrian Calborean, George Mile, Mihai Vidrighin, Monica Iosin, Loredana Leopold, Dana Maniu, Nicolae Leopold, Vasile Chiş
ABSTRACT
Absorption spectra of PTCDI (3,4,9,10-perylene-tetracarboxylic-diimide) have been investigated in chloroform, N,N'-dimethylformamide (DMF) and dimethylsulfoxide (DMSO). While no signature of assembled PTCDI molecules is observed in chloroform solution, distinct bands assigned to their aggregation have been identified in DMF and DMSO solutions. PTCDI monomers show very similar absorption patterns in chloroform and DMSO solutions. Experimental data, including the vibronic structure of the absorption spectra were explained based on the Franck-Condon approximation and quantum chemical results obtained at PBE0-DCP/6-31+G(d,p) level of theory. Geometry optimization of the first excited state leads to a nice agreement between the calculated adiabatic transition energies and experimental data.
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%