Merck
  • Kinetic resolution of propargylamines via a highly enantioselective non-enzymatic N-acylation process.
Kinetic resolution of propargylamines via a highly enantioselective non-enzymatic N-acylation process.
Chemical communications (Cambridge, England) (2012-09-22)
Amandine Kolleth, Sarah Christoph, Stellios Arseniyadis, Janine Cossy
ABSTRACT
The non-enzymatic kinetic resolution of diversely substituted primary propargylic amines is reported featuring a highly selective acetyl transfer using (1S,2S)- in conjunction with Aliquat(TM) 336, affording the corresponding enantio-enriched N-acetylated propargylic amines with unprecedented levels of selectivity (s-factors of up to 193 at 50% conversion).
Sigma-Aldrich
Propargylamine hydrochloride, 95%
Sigma-Aldrich
N-Methyl-N-propargylbenzylamine, 97%
Sigma-Aldrich
Pargyline hydrochloride
Sigma-Aldrich
Propargylamine, 98%