Merck
  • Substrate stiffness regulates cadherin-dependent collective migration through myosin-II contractility.
Substrate stiffness regulates cadherin-dependent collective migration through myosin-II contractility.
The Journal of cell biology (2012-10-24)
Mei Rosa Ng, Achim Besser, Gaudenz Danuser, Joan S Brugge
ABSTRACT
The mechanical microenvironment is known to influence single-cell migration; however, the extent to which mechanical cues affect collective migration of adherent cells is not well understood. We measured the effects of varying substrate compliance on individual cell migratory properties in an epithelial wound-healing assay. Increasing substrate stiffness increased collective cell migration speed, persistence, and directionality as well as the coordination of cell movements. Dynamic analysis revealed that wounding initiated a wave of motion coordination from the wound edge into the sheet. This was accompanied by a front-to-back gradient of myosin-II activation and establishment of cell polarity. The propagation was faster and farther reaching on stiff substrates, indicating that substrate stiffness affects the transmission of directional cues. Manipulation of myosin-II activity and cadherin-catenin complexes revealed that this transmission is mediated by coupling of contractile forces between neighboring cells. Thus, our findings suggest that the mechanical environment integrates in a feedback with cell contractility and cell-cell adhesion to regulate collective migration.
Sigma-Aldrich
Anti-Vinculin antibody, Mouse monoclonal, clone hVIN-1, purified from hybridoma cell culture
Sigma-Aldrich
Polyacrylamide, average Mn 150,000
Sigma-Aldrich
Polyacrylamide, nonionic water-soluble polymer